Dibenzyl diisopropylphosphoramidite ≥95%

Supplier: BeanTown Chemical

Synonyms: Dibenzyl N,N-diisopropylphosphoramidite

225405-100G 225405-25G 225405-5G
BT225405-100GEA 1022 USD
BT225405-100G BT225405-25G BT225405-5G
Dibenzyl diisopropylphosphoramidite ≥95%
Dibenzyl diisopropylphosphoramidite

CAS: 108549-23-1; MDL No: MFCD00191988 Liquid; Molecular Formula: C20H28NO2P; MW: 345.43 Boiling Point: 130°/0.55 mmHg; Flash point: 69°C (156°F) Density (g/mL): 1.028; Refractive Index: 1.535 Moisture Sensitive


  • Refractive index: 1.535
  • Moisture Sensitive
  • Liquid

Formula: C₂₀H₂₈NO₂P
MW: 345.43 g/mol
Boiling Pt: 130 °C/0.55 mmHg
Density: 1.028 g/mL
Flash Pt: 69 °C (156 °F)
MDL Number: MFCD00191988
CAS Number: 108549-23-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR