4,4-Dimethylcyclohexanone ≥98%
Supplier: BeanTown Chemical
Synonyms:
4,4-Dimethylcyclohexan-1-one
225380-1G
225380-250MG
225380-5G
BT225380-1GEA
95
USD
BT225380-1G
BT225380-250MG
BT225380-5G
4,4-Dimethylcyclohexanone ≥98%
4,4-Dimethylcyclohexanone
CAS: 4255-62-3; MDL No: MFCD00234965
Solid; Molecular Formula: C8H14O; MW: 126.20
Melting Point: 41-43°; Flash point: 62°C (144°F)
- Solid
Formula:
C₈H₁₄O Melting Pt: 41…43 °C Flash Pt: 62 °C (144 °F) |
MDL Number:
MFCD00234965 CAS Number: 4255-62-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...