3-Biphenylboronic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: [1,1'-Biphenyl]-3-ylboronic acid

225370-1G 225370-5G
BT225370-1GEA 39 USD
BT225370-1G BT225370-5G
3-Biphenylboronic acid ≥98%
3-Biphenylboronic acid

CAS: 5122-95-2; MDL No: MFCD01318102 Powder; Molecular Formula: C12H11BO2; MW: 198.03 Melting Point: 193-198°


  • Powder

Formula: C₁₂H₁₁BO₂
MW: 198.03 g/mol
Melting Pt: 193…198 °C
MDL Number: MFCD01318102
CAS Number: 5122-95-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR