Silver diethyldithiocarbamate ACS
Supplier: BeanTown Chemical
Synonyms:
N,N-Diethyldithiocarbamic acid silver salt, Diethyldithiocarbamic acid silver salt, DETC, (Diethyldithiocarbamato-S,S')silver
225335-10G
225335-50G
BT225335-10GEA
202.06
USD
BT225335-10G
BT225335-50G
Silver diethyldithiocarbamate ACS
Silver diethyldithiocarbamate
CAS: 1470-61-7; EC No: 216-003-7; MDL No: MFCD00004929
Powder; Linear Formula: (CH3CH2)2NCSSAg; Molecular Formula: C5H10AgNS2; MW: 256.14
Melting Point: 172-175°
Light Sensitive
- Light Sensitive
- Powder
Formula:
(C₂H₅)₂NCSSAg MW: 256.14 g/mol Melting Pt: 172…175 °C |
MDL Number:
MFCD00004929 CAS Number: 1470-61-7 EINECS: 216-003-7 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...