Bromotrispyrrolidinophosphonium hexafluorophosphate ≥97%

Supplier: BeanTown Chemical

Synonyms: Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, PyBrOP , Bromotripyrrolidinylphosphonium hexafluorophosphate, Bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)

225320-1G 225320-5G
BT225320-1GEA 44 USD
BT225320-1G BT225320-5G
Bromotrispyrrolidinophosphonium hexafluorophosphate ≥97%
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate

CAS: 132705-51-2; MDL No: MFCD00077412 Powder; Molecular Formula: C12H24BrF6N3P2; MW: 466.18 Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₁₂H₂₄BrF₆N₃P₂
MW: 466.2 g/mol
MDL Number: MFCD00077412
CAS Number: 132705-51-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR