Lecithin (from soy bean)
Supplier: BeanTown Chemical
225315-1KG
225315-250G
BT225315-1KGEA
156.68
USD
BT225315-1KG
BT225315-250G
Lecithin (from soy bean)
Lecithin
CAS: 8002-43-5; EC No: 232-307-2; MDL No: MFCD00147406; RTECS: OG7565000
Solid
- RTECS: OG7565000
- Solid
MDL Number:
MFCD00147406 CAS Number: 8002-43-5 EINECS: 232-307-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...