3-Nitroisonicotinic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-Nitropyridine-4-carboxylic acid

225290-1G 225290-250MG
BT225290-1GEA 202 USD
BT225290-1G BT225290-250MG
3-Nitroisonicotinic acid ≥97%
3-Nitroisonicotinic acid

CAS: 59290-82-3; MDL No: MFCD04114244 Powder; Molecular Formula: C6H4N2O4; MW: 168.11


  • Powder

Formula: C6H4N2O4
MW: 168.11 g/mol
MDL Number: MFCD04114244
CAS Number: 59290-82-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR