4'-Methoxychalcone ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-(4-Methoxyphenyl)-3-phenylprop-2-en-1-one, α-Styryl p-anisyl ketone, 4'-Methoxybenzylidenacetophenone

225285-100G 225285-25G 225285-5G
BT225285-100GEA 228 USD
BT225285-100G BT225285-25G BT225285-5G
4'-Methoxychalcone ≥97%
4'-Methoxychalcone

CAS: 959-23-9; EC No: 213-495-5; MDL No: MFCD00008407 Crystalline/Powder; Molecular Formula: C16H14O2 ; MW: 238.29 Melting Point: 100-103°


  • Crystalline/Powder

Formula: C₁₆H₁₄O₂
MW: 238.29 g/mol
Melting Pt: 100…103 °C
MDL Number: MFCD00008407
CAS Number: 959-23-9
EINECS: 213-495-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR