Trimethyl phosphonoacetate ≥98%

Supplier: BeanTown Chemical

Synonyms: Methyl 2-(dimethoxyphosphoryl)acetate, Dimethyl methoxycarbonylmethanephosphonate

225280-100G 225280-25G 225280-500G
BT225280-100GEA 124.2 USD
BT225280-100G BT225280-25G BT225280-500G
Trimethyl phosphonoacetate ≥98%
Trimethyl phosphonoacetate

CAS: 5927-18-4; EC No: 227-663-0; MDL No: MFCD00008452 Liquid; Linear Formula: (CH3O)2P(O)CH2COOCH3; Molecular Formula: C5H11O5P; MW: 182.11 Boiling Point: 265-268°; Flash point: 70°C (158°F) Density (g/mL): 1.125; Refractive Index: 1.437


  • Refractive index: 1.437
  • Liquid

Formula: C₅H₁₁O₅P
MW: 182.11 g/mol
Boiling Pt: 265…268 °C
Density: 1.125 g/mL
Flash Pt: 70 °C (158 °F)
MDL Number: MFCD00008452
CAS Number: 5927-18-4
EINECS: 227-663-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR