Polyvinyl alcohol 98-99%, high molecular weight, hydrolysed
Supplier: BeanTown Chemical
Synonyms:
Poly(vinyl alcohol), PVOH, PVA, Mowiol®
225255-100G
225255-25G
225255-500G
BT225255-100GEA
32.99
USD
BT225255-100G
BT225255-25G
BT225255-500G
Polyvinyl alcohol 98-99%, high molecular weight, hydrolysed
Polyvinyl alcohol
CAS: 9002-89-5; MDL No: MFCD00081922
Crystalline; Linear Formula: [-CH2CH(OH)-]n
Melting Point: 200°
Density (g/mL): 1.269
- Crystalline
Formula:
[-CH₂CH(OH)-]n Melting Pt: 200 °C Density: 1.269 g/mL |
MDL Number:
MFCD00081922 CAS Number: 9002-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...