Ethyl-5-hydroxyindole-2-carboxylate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl 5-hydroxyindole-2-carboxylate

225245-1G 225245-250MG 225245-5G
BT225245-1GEA 145 USD
BT225245-1G BT225245-250MG BT225245-5G
Ethyl-5-hydroxyindole-2-carboxylate ≥98%
Ethyl-5-hydroxyindole-2-carboxylate

CAS: 24985-85-1; EC No: 246-554-9; MDL No: MFCD00015458 Crystalline/Powder; Molecular Formula: C11H11NO3; MW: 205.21 Melting Point: 150-151°


  • Crystalline/Powder

Formula: C₁₁H₁₁NO₃
MW: 205.213 g/mol
Melting Pt: 150…151 °C
MDL Number: MFCD00015458
CAS Number: 24985-85-1
EINECS: 246-554-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR