Picolinic acid ≥99%
Supplier: BeanTown Chemical
225240-100G
225240-500G
BT225240-100GEA
45.2
USD
BT225240-100G
BT225240-500G
Picolinic acid ≥99%
Picolinic acid
CAS: 98-98-6; EC No: 202-719-7; MDL No: MFCD00006293; RTECS: TJ7344000
Crystalline; Molecular Formula: C6H5NO2 ; MW: 123.11
Melting Point: 139-142°
- RTECS: TJ7344000
- Crystalline
Formula:
C₆H₅NO₂ Melting Pt: 139…142 °C |
MDL Number:
MFCD00006293 CAS Number: 98-98-6 EINECS: 202-719-7 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...