(1R)-(+)-2,10,Camphorsultam ≥99%
Supplier: BeanTown Chemical
Synonyms:
Ticagrelor Impurity 113, (3aR)-8,8-dimethylhexahydro-2H-3a,6-methanobenzo[d]isothiazole 1,1-dioxide, (3aR,6S,7aS)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide
225235-1G
225235-5G
BT225235-1GEA
79
USD
BT225235-1G
BT225235-5G
(1R)-(+)-2,10,Camphorsultam ≥99%
(1R)-(+)-2,10,Camphorsultam
CAS: 108448-77-7; MDL No: MFCD00151762
Crystalline; Molecular Formula: C10H17NO2S ; MW: 215.31
Boiling Point: 185-187°
Optical Rotation: [α]20/D +32°, c = 5 in chloroform
- Optical rotation: [α]20/D +32°, c = 5 in chloroform
- Crystalline
Formula:
C₁₀H₁₇NO₂S MW: 215.32 g/mol Boiling Pt: 185…187 °C |
MDL Number:
MFCD00151762 CAS Number: 108448-77-7 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...