L-Theanine ≥99%

Supplier: BeanTown Chemical

225225-1G 225225-5G
BT225225-1GEA 105 USD
BT225225-1G BT225225-5G
L-Theanine ≥99%
L-Theanine

CAS: 3081-61-6; EC No: 221-379-0; MDL No: MFCD00059653 Solid; Molecular Formula: C7H14N2O3; MW: 174.20 Density (g/mL): 1.174


  • Solid

Formula: C₇H₁₄N₂O₃
Density: 1.174 g/mL
MDL Number: MFCD00059653
CAS Number: 3081-61-6
EINECS: 221-379-0

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR