1-Acetamidoadamantane ≥98%

Supplier: BeanTown Chemical

Synonyms: N-1-Adamantylacetamide, N-Acetyl-1-adamantanamine

225180-10G 225180-50G
BT225180-10GEA 26.61 USD
BT225180-10G BT225180-50G
1-Acetamidoadamantane ≥98%
1-Acetamidoadamantane

CAS: 880-52-4; EC No: 212-914-9; MDL No: MFCD00074730; RTECS: AB4308000 Powder; Molecular Formula: C12H19NO; MW: 193.29


  • RTECS: AB4308000
  • Powder

Formula: C₁₂H₁₉NO
MW: 193.29 g/mol
MDL Number: MFCD00074730
CAS Number: 880-52-4
EINECS: 212-914-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR