4-Methyl-3-(trifluoromethyl)benzoic acid ≥97%
Supplier: BeanTown Chemical
225155-1G
225155-5G
BT225155-1GEA
83
USD
BT225155-1G
BT225155-5G
4-Methyl-3-(trifluoromethyl)benzoic acid ≥97%
4-Methyl-3-(trifluoromethyl)benzoic acid
CAS: 261952-01-6; MDL No: MFCD01631592
Crystalline; Molecular Formula: C9H7F3O2; MW: 204.15
Melting Point: 175-179°
- Crystalline
Formula:
C₉H₇F₃O₂ Melting Pt: 175…179 °C |
MDL Number:
MFCD01631592 CAS Number: 261952-01-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...