Methylsulfonylacetonitrile ≥98%

Supplier: BeanTown Chemical

225140-10G 225140-50G
BT225140-10GEA 131 USD
BT225140-10G BT225140-50G
Methylsulfonylacetonitrile ≥98%
Methylsulfonylacetonitrile

CAS: 2274-42-2; EC No: 218-888-5; MDL No: MFCD00014743 Solid; Molecular Formula: C3H5NO2S; MW: 119.14 Melting Point: 81-84°


  • Solid

Formula: C₃H₅NO₂S
Melting Pt: 81…84 °C
MDL Number: MFCD00014743
CAS Number: 2274-42-2
EINECS: 218-888-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR