Diethylene glycol monobutyl ether ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-(2-Butoxyethoxy)ethanol, Diethylene glycol mono-n-butyl ether, Butyldigol

225115-1L 225115-5L
BT225115-1LEA 60.35 USD
BT225115-1L BT225115-5L
Diethylene glycol monobutyl ether ≥99%
Diethylene glycol monobutyl ether

CAS: 112-34-5; EC No: 203-961-6; MDL No: MFCD00002881; RTECS: KJ9100000 Liquid; Linear Formula: CH3(CH2)3OCH2CH2OCH2CH2OH; Molecular Formula: C8H18O3; MW: 162.23 Melting Point: -68°; Boiling Point: 231°; Flash point: 99°C (210°F) Density (g/mL): 0.967; Refractive Index: 1.432


  • RTECS: KJ9100000
  • Refractive index: 1.432
  • Liquid

Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MW: 162.23 g/mol
Boiling Pt: 230.4
Melting Pt: -68
Density: 0.967 g/mL
Flash Pt: 99 °C (210 °F)
MDL Number: MFCD00002881
CAS Number: 112-34-5
EINECS: 203-961-6
Merck Index: 13,01556

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR