Tin ≥99.9%, Powder -100 mesh

Supplier: BeanTown Chemical

225085-100G 225085-500G
BT225085-100GEA 63.81 USD
BT225085-100G BT225085-500G
Tin ≥99.9%, Powder -100 mesh
Tin

CAS: 7440-31-5; EC No: 231-141-8; MDL No: MFCD00133862 Powder; Molecular Formula: Sn; MW: 118.71 Melting Point: 231.9°; Boiling Point: 2270° Density (g/mL): 7.31


  • Powder

Formula: Sn
Boiling Pt: 2270 °C
Melting Pt: 231.9 °C
Density: 7.31 g/mL
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR