trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Fluoro-4'-(trans-4-propylcyclohexyl)biphenyl, 4-Fluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

225080-1G 225080-5G
BT225080-1GEA 75 USD
BT225080-1G BT225080-5G
trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl ≥97%
trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl

CAS: 87260-24-0; MDL No: MFCD11053438 Solid; Molecular Formula: C21H25F; MW: 296.43 Melting Point: 104-106°


  • Solid

Formula: C₂₁H₂₅F
Melting Pt: 104…106 °C
MDL Number: MFCD11053438
CAS Number: 87260-24-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR