2,4-Dichloro-5-methylpyridine

Supplier: BeanTown Chemical

Synonyms: 2,4-Dichloro-5-picoline

225075-1G 225075-250MG
BT225075-1GEA 130 USD
BT225075-1G BT225075-250MG
2,4-Dichloro-5-methylpyridine
2,4-Dichloro-5-methylpyridine

CAS: 56961-78-5 Liquid; Molecular Formula: C6H5Cl2N ; MW: 162.02


  • Liquid

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
MDL Number: MFCD08704813
CAS Number: 56961-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR