Potassium dihydrogen phosphate ≥99.0% ACS
Supplier: BeanTown Chemical
225070-1KG
225070-500G
BT225070-1KGEA
127.36
USD
BT225070-1KG
BT225070-500G
Potassium dihydrogen phosphate ≥99.0% ACS
Potassium dihydrogen phosphate
CAS: 7778-77-0; EC No: 231-913-4; MDL No: MFCD00011401; RTECS: TC6615500
Crystalline; Linear Formula: KH2PO4; MW: 136.09
Melting Point: 253°
Density (g/mL): 2.338
Hygroscopic
- RTECS: TC6615500
- Hygroscopic
- Crystalline
Formula:
KH₂PO₄ Melting Pt: 253 °C Density: 2.338 g/mL |
MDL Number:
MFCD00011401 CAS Number: 7778-77-0 EINECS: 231-913-4 |
Specification Test Results
Assay | 99.0% min. |
pH of 5% solution at 25°C | 4.1-4.5 |
Insoluble matter | ≤0.01% |
Loss on drying (105°C) | ≤0.2% |
Chloride (Cl) | ≤0.001% |
Sulfate (SO4) | ≤0.003% |
Heavy metals (as Pb) | ≤0.001% |
Iron (Fe) | ≤0.002% |
Sodium (Na) | ≤0.005% |
Nitrogen compounds (as N) | ≤0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...