2-Amino-4,6-dimethoxypyrimidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 4,6-Dimethoxypyrimidin-2-amine, 4,6-Dimethoxypyrimidin-2-ylamine

225050-25G 225050-5G
BT225050-25GEA 156 USD
BT225050-25G BT225050-5G
2-Amino-4,6-dimethoxypyrimidine ≥98%
2-Amino-4,6-dimethoxypyrimidine

CAS: 36315-01-2; EC No: 252-969-6; MDL No: MFCD00038832 Solid; Molecular Formula: C6H9N3O2; MW: 155.15 Melting Point: 94-96°


  • Solid

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD00038832
CAS Number: 36315-01-2
EINECS: 252-969-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR