p-Anisic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Methoxybenzoic acid, 4-Anisic acid
225030-1KG
225030-250G
225030-5KG
BT225030-1KGEA
148
USD
BT225030-1KG
BT225030-250G
BT225030-5KG
p-Anisic acid ≥98%
p-Anisic acid
CAS: 100-09-4; EC No: 202-818-5; MDL No: MFCD00002542; RTECS: BZ4395000
Powder; Molecular Formula: C8H8O3; MW: 152.15
Melting Point: 182-185°
- RTECS: BZ4395000
- Powder
Formula:
C₈H₈O₃ MW: 152.15 g/mol Melting Pt: 182…185 °C |
MDL Number:
MFCD00002542 CAS Number: 100-09-4 EINECS: 202-818-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...