Magnesium titanate 99

Supplier: BeanTown Chemical

225015-1KG 225015-250G
BT225015-1KGEA 83 USD
BT225015-1KG BT225015-250G
Magnesium titanate 99
Magnesium titanate

CAS: 12032-30-3; EC No: 234-766-4; MDL No: MFCD00016198 2 Micron Powder; Molecular Formula: MgO3Ti; MW: 120.18 Melting Point: 1610°


  • 2 Micron Powder

Formula: MgO₃Ti
MW: 120.18
Melting Pt: 1610 °C
MDL Number: MFCD00016198
CAS Number: 12032-30-3
EINECS: 234-766-4

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR