Chromium ≥99%, irregular pieces

Supplier: BeanTown Chemical

224980-100G 224980-500G
BT224980-100GEA 46 USD
BT224980-100G BT224980-500G
Chromium ≥99%, irregular pieces
Chromium

CAS: 7440-47-3; EC No: 231-157-5; MDL No: MFCD00010944 Solid; Molecular Formula: Cr; MW: 52.00 Melting Point: 1857°; Boiling Point: 2672° Density (g/mL): 7.14


  • Solid

Formula: Cr
MW: 52
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.14 g/mL
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR