3-Fluoroanthranilic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Amino-3-fluorobenzoic acid
224970-25G
224970-5G
BT224970-25GEA
105
USD
BT224970-25G
BT224970-5G
3-Fluoroanthranilic acid ≥98%
3-Fluoroanthranilic acid
CAS: 825-22-9; MDL No: MFCD01569395
Solid; Linear Formula: FC6H4(NH2)(COOH); Molecular Formula: C7H6FNO2; MW: 155.13
Melting Point: 171-172°
- Solid
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Melting Pt: 171…172 °C |
MDL Number:
MFCD01569395 CAS Number: 825-22-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...