Cobalt iron oxide

Supplier: BeanTown Chemical
Danger

224965-25G 224965-5G
BT224965-25GEA 180 USD
BT224965-25G BT224965-5G
Cobalt iron oxide
Cobalt iron oxide

CAS: 12052-28-7; MDL No: MFCD00016018 Powder; Molecular Formula: CoFe2O4; MW: 234.62


  • Powder

Formula: CoFe₂O₄
MDL Number: MFCD00016018
CAS Number: 12052-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR