2-Chloro-3-nitropyridine ≥99%

Supplier: BeanTown Chemical

224960-100G 224960-25G 224960-5G
BT224960-100GEA 385 USD
BT224960-100G BT224960-25G BT224960-5G
2-Chloro-3-nitropyridine ≥99%
2-Chloro-3-nitropyridine

CAS: 5470-18-8; EC No: 226-799-8; MDL No: MFCD00006232 Powder; Molecular Formula: C5H3ClN2O2 ; MW: 158.54 Melting Point: 100-103°; Flash point: 185°C (365°F)


  • Powder

Formula: C₅H₃ClN₂O₂
Melting Pt: 100…103 °C
Flash Pt: 185 °C (365 °F)
MDL Number: MFCD00006232
CAS Number: 5470-18-8
EINECS: 226-799-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR