3-Fluorosalicylaldehyde ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-Fluoro-2-hydroxybenzaldehyde
224955-1G
224955-250MG
224955-5G
BT224955-1GEA
131
USD
BT224955-1G
BT224955-250MG
BT224955-5G
3-Fluorosalicylaldehyde ≥98%
3-Fluorosalicylaldehyde
CAS: 394-50-3; MDL No: MFCD00003319
Powder; Molecular Formula: C7H5FO2; MW: 140.11
Melting Point: 68-70°
Air Sensitive
- Air Sensitive
- Powder
Formula:
C₇H₅FO₂ MW: 140.12 g/mol Melting Pt: 68…70 °C |
MDL Number:
MFCD00003319 CAS Number: 394-50-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...