Ammonium carbamate ≥98%

Supplier: BeanTown Chemical

Synonyms: Carbamic acid ammonium salt

224945-250G 224945-50G
BT224945-250GEA 134.84 USD
BT224945-250G BT224945-50G
Ammonium carbamate ≥98%
Ammonium carbamate

CAS: 1111-78-0; EC No: 214-185-2; MDL No: MFCD00013010; RTECS: EY8575000 Powder; Linear Formula: NH2CO2NH4; Molecular Formula: CH6N2O2; MW: 78.07


  • RTECS: EY8575000
  • Powder

Formula: H₂NCO₂NH₄
MW: 78.07 g/mol
MDL Number: MFCD00013010
CAS Number: 1111-78-0
EINECS: 214-185-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR