D-(-)-Leucine ≥99%

Supplier: BeanTown Chemical

Synonyms: H-D-Leu-OH, (R)-(-)-Leucine

224935-100G 224935-25G 224935-5G
BT224935-100GEA 660 USD
BT224935-100G BT224935-25G BT224935-5G
D-(-)-Leucine ≥99%
D-(-)-Leucine

CAS: 328-38-1; EC No: 206-327-7; MDL No: MFCD00063088; RTECS: OH2840000 Crystalline; Molecular Formula: C6H13NO2; MW: 131.17 Optical Rotation: [α]23/D -14.7°, c = 2 in 5M HCl


  • RTECS: OH2840000
  • Optical rotation: [α]23/D -14.7°, c = 2 in 5M HCl
  • Crystalline

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
MDL Number: MFCD00063088
CAS Number: 328-38-1
EINECS: 206-327-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR