1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose ≥98%

Supplier: BeanTown Chemical

Synonyms: Diacetone-D-glucose, (-)-1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose, (3aR,5S,6S,6aR)-5-((R)-2,2-Dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol, D-Glucose diacetonide

224930-100G 224930-25G 224930-500G
BT224930-100GEA 142 USD
BT224930-100G BT224930-25G BT224930-500G
1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose ≥98%
1,2:5,6-Di-O-isopropylidene-alpha-D-glucofuranose

CAS: 582-52-5; EC No: 209-486-0; MDL No: MFCD00005544; RTECS: LZ4958000 Crystalline/Powder; Molecular Formula: C12H20O6; MW: 260.28 Melting Point: 109-113° Optical Rotation: [α]20/D -18°, c = 1% in water


  • RTECS: LZ4958000
  • Optical rotation: [α]20/D -18°, c = 1% in water
  • Crystalline/Powder

Formula: C₁₂H₂₀O₆
MW: 260.29 g/mol
Melting Pt: 109…113 °C
MDL Number: MFCD00005544
CAS Number: 582-52-5
EINECS: 209-486-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR