Germanium ≥99.999% (trace metals basis), pieces ≤3.2 mm
Supplier: BeanTown Chemical
224910-10G
224910-50G
BT224910-10GEA
203
USD
BT224910-10G
BT224910-50G
Germanium ≥99.999% (trace metals basis), pieces ≤3.2 mm
Germanium
CAS: 7440-56-4; EC No: 231-164-3; MDL No: MFCD00085310; RTECS: LY5200000
Solid; Molecular Formula: Ge; MW: 72.64
Melting Point: 937°; Boiling Point: 2830°
Density (g/mL): 5.35
- RTECS: LY5200000
- Solid
Formula:
Ge Boiling Pt: 2830 °C Melting Pt: 937 °C Density: 5.35 g/mL |
MDL Number:
MFCD00085310 CAS Number: 7440-56-4 EINECS: 231-164-3 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...