4-Chloro-6-methoxyquinoline
Supplier: BeanTown Chemical
224835-1G
224835-5G
BT224835-1GEA
123
USD
BT224835-1G
BT224835-5G
4-Chloro-6-methoxyquinoline
4-Chloro-6-methoxyquinoline
CAS: 4295-04-9; MDL No: MFCD00024007
Solid; Molecular Formula: C10H8ClNO; MW: 193.63
- Solid
Formula:
C₁₀H₈ClNO MW: 193.6300 g/mol |
MDL Number:
MFCD00024007 CAS Number: 4295-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...