exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione
Supplier: BeanTown Chemical
224790-1G
224790-5G
BT224790-1GEA
74
USD
BT224790-1G
BT224790-5G
exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione
exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione
CAS: 14805-29-9
Solid; Molecular Formula: C9H11NO2; MW: 165.191
- Solid
Formula:
C₉H₁₁NO₂ |
MDL Number:
MFCD12964181 CAS Number: 14805-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...