Ranolazine

Supplier: BeanTown Chemical

224750-1G 224750-5G
BT224750-1GEA 37 USD
BT224750-1G BT224750-5G
Ranolazine
Ranolazine

CAS: 95635-55-5; MDL No: MFCD03788770 Solid; Molecular Formula: C24H33N3O4; MW: 427.54


  • Solid

Formula: C₂₄H₃₃N₃O₄
MDL Number: MFCD00864690
CAS Number: 95635-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR