2-Chloro-4-hydrazinopyridine ≥95%

Supplier: BeanTown Chemical

224725-1G 224725-5G
BT224725-1GEA 135 USD
BT224725-1G BT224725-5G
2-Chloro-4-hydrazinopyridine ≥95%
2-Chloro-4-hydrazinopyridine

CAS: 700811-29-6 Solid; Linear Formula: ClC6H3NNHNH2; Molecular Formula: C5H6ClN3; MW: 143.57


  • Solid

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD08460247
CAS Number: 700811-29-6

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR