DL-α-Phenylalanine ≥99%
Supplier: BeanTown Chemical
Synonyms:
H-DL-Phe-OH, DL-Phenylalanine, 3-Phenyl-DL-alanine
224715-100G
224715-25G
224715-500G
BT224715-100GEA
52.7
USD
BT224715-100G
BT224715-25G
BT224715-500G
DL-α-Phenylalanine ≥99%
DL-α-Phenylalanine
CAS: 150-30-1; EC No: 205-756-7; MDL No: MFCD00064225
Powder; Molecular Formula: C9H11NO2; MW: 165.19
Melting Point: 266-267° (decomposes)
- Powder
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol Melting Pt: 266…267 °C (dec.) |
MDL Number:
MFCD00064225 CAS Number: 150-30-1 EINECS: 205-756-7 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...