Oleyl alcohol 80-85%, Technical Grade
Supplier: BeanTown Chemical
224635-100G
224635-500G
BT224635-100GEA
29.34
USD
BT224635-100G
BT224635-500G
Oleyl alcohol 80-85%, Technical Grade
Oleyl alcohol
CAS: 143-28-2; EC No: 205-597-3; MDL No: MFCD00002993; RTECS: RG4120000
Liquid; Linear Formula: CH3(CH2)7CH=CH(CH2)7CH2OH; Molecular Formula: C18H36O; MW: 268.48
Melting Point: 0-5°; Boiling Point: 206-207°/13 mmHg; Flash point: 170°C (338°F)
Density (g/mL): 0.849; Refractive Index: 1.46
- RTECS: RG4120000
- Refractive index: 1.46
- Liquid
Formula:
C₁₈H₃₆O MW: 268.48 g/mol Boiling Pt: 206…207 °C/13 mmHg Melting Pt: 0…5 °C Density: 0.849 g/mL Flash Pt: 170 °C (338 °F) |
MDL Number:
MFCD00002993 CAS Number: 143-28-2 EINECS: 205-597-3 |
Specification Test Results
Assay | 80-85% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...