(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) ≥98%

Supplier: BeanTown Chemical

Synonyms: o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

224630-1G 224630-5G
BT224630-1GEA 43.73 USD
BT224630-1G BT224630-5G
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) ≥98%
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG)

CAS: 369-07-3; EC No: 206-716-1; MDL No: MFCD00063255 Solid; Molecular Formula: C12H15NO8; MW: 301.25 Optical Rotation: [α]20/D -70±1°, c = 1% in water


  • Optical rotation: [α]20/D -70±1°, c = 1% in water
  • Solid

Formula: C₁₂H₁₅NO₈
MDL Number: MFCD00063255
CAS Number: 369-07-3
EINECS: 206-716-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR