3-Amino-4-iodopyridine ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Iodo-3-pyridylamine, 4-Iodopyridin-3-amine, 4-Iodo-pyridin-3-ylamine
224615-1G
224615-5G
BT224615-1GEA
240
USD
BT224615-1G
BT224615-5G
3-Amino-4-iodopyridine ≥98%
3-Amino-4-iodopyridine
CAS: 105752-11-2; MDL No: MFCD04971334
Solid; Molecular Formula: C5H5IN2; MW: 220.01
Melting Point: 65-70°
Air Sensitive, Light Sensitive
- Air Sensitive, Light Sensitive
- Solid
Formula:
C₅H₅IN₂ Melting Pt: 65…70 °C |
MDL Number:
MFCD04971334 CAS Number: 105752-11-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...