Cyclohexyldiphenylphosphine ≥98%

Supplier: BeanTown Chemical

Synonyms: Diphenylcyclohexylphosphine

224585-1G 224585-25G 224585-5G
BT224585-1GEA 34 USD
BT224585-1G BT224585-25G BT224585-5G
Cyclohexyldiphenylphosphine ≥98%
Cyclohexyldiphenylphosphine

CAS: 6372-42-5; EC No: 228-904-2; MDL No: MFCD00046360; RTECS: SY8387500 Solid; Molecular Formula: C18H21P ; MW: 268.33 Melting Point: 58-62°; Flash point: 110°C (230°F) Air Sensitive


  • RTECS: SY8387500
  • Air Sensitive
  • Solid

Formula: C₁₈H₂₁P
MW: 268.33 g/mol
Melting Pt: 58…62 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00046360
CAS Number: 6372-42-5
EINECS: 228-904-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR