4-Bromostyrene ≥98% stabilized

Supplier: BeanTown Chemical

Synonyms: 1-Bromo-4-vinylbenzene

224565-100G 224565-25G 224565-5G
BT224565-100GEA 523 USD
BT224565-100G BT224565-25G BT224565-5G
4-Bromostyrene ≥98% stabilized
4-Bromostyrene

CAS: 2039-82-9; EC No: 218-022-6; MDL No: MFCD00000110 Liquid; Molecular Formula: C8H7Br ; MW: 183.05 Boiling Point: 89°/16 mmHg; Flash point: 75°C (167°F) Density (g/mL): 1.4; Refractive Index: 1.594


  • Refractive index: 1.594
  • Liquid

Formula: C₈H₇Br
MW: 183.05 g/mol
Boiling Pt: 89 °C/16 mmHg
Density: 1.4 g/mL
Flash Pt: 75 °C (167 °F)
MDL Number: MFCD00000110
CAS Number: 2039-82-9
EINECS: 218-022-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR