Iron ≥99.99% (trace metals basis), wire Ø 1.0 mm
Supplier: BeanTown Chemical
224560-250CM
224560-50CM
BT224560-250CMEA
393.3
USD
BT224560-250CM
BT224560-50CM
Iron ≥99.99% (trace metals basis), wire Ø 1.0 mm
Iron
CAS: 7439-89-6; EC No: 231-096-4; MDL No: MFCD00010999
Solid; Molecular Formula: Fe; MW: 55.85
Melting Point: 1535°; Boiling Point: 2750°
Density (g/mL): 7.86
- Solid
Formula:
Fe Boiling Pt: 2750 °C Melting Pt: 1535 °C Density: 7.86 g/mL |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...