Diisopropyl phosphonate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Diisopropyl phosphite, Di-i-propylphosphite, Diisopropyl hydrogen phosphite
224525-100G
224525-25G
BT224525-100GEA
118
USD
BT224525-100G
BT224525-25G
Diisopropyl phosphonate ≥98%
Diisopropyl phosphonate
CAS: 1809-20-7; EC No: 217-317-7; MDL No: MFCD00117905; RTECS: SZ7660000
Liquid; Molecular Formula: C6H15O3P; MW: 166.16
Boiling Point: 72-75°/10 mmHg
Density (g/mL): 0.990
Moisture Sensitive
- RTECS: SZ7660000
- Moisture Sensitive
- Liquid
Formula:
C₆H₁₅O₃P MW: 166.16 g/mol Boiling Pt: 72…75 °C/10 mmHg Density: 0.990 g/mL |
MDL Number:
MFCD00117905 CAS Number: 1809-20-7 EINECS: 217-317-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...