trans-2-Aminocyclohexanol hydrochloride ≥99%

Supplier: BeanTown Chemical

224495-100G 224495-25G 224495-5G
BT224495-100GEA 890 USD
BT224495-100G BT224495-25G BT224495-5G
trans-2-Aminocyclohexanol hydrochloride ≥99%
trans-2-Aminocyclohexanol hydrochloride

CAS: 5456-63-3; MDL No: MFCD00001490 Powder; Linear Formula: H2NC6H4OH·HCl; Molecular Formula: C6H13NO·HCl; MW: 151.64 Melting Point: 172-175° Hygroscopic


  • Hygroscopic
  • Powder

Formula: C₆H₁₄ClNO
Melting Pt: 172…175 °C
MDL Number: MFCD00001490
CAS Number: 5456-63-3

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR