N,N-Dimethylacetamide ≥99.5% for HPLC
Supplier: BeanTown Chemical
Synonyms:
Acetic acid dimethylamide, Dimethyl Acetamide, DMAC
224485-1L
BT224485-1LEA
104.27
USD
BT224485-1L
BT224485-4L
BT224485-4X1L
N,N-Dimethylacetamide ≥99.5% for HPLC
N,N-Dimethylacetamide
CAS: 127-19-5; EC No: 204-826-4; MDL No: MFCD00008686; RTECS: AB7700000
Liquid; Linear Formula: CH2CON(CH3)2; Molecular Formula: C4H9NO ; MW: 87.12
Melting Point: -20°; Boiling Point: 165-166°; Flash point: 70°C (158°F)
Density (g/mL): 0.937; Refractive Index: 1.437
Hygroscopic
- RTECS: AB7700000
- Refractive index: 1.437
- Hygroscopic
- Liquid
Formula:
CH₃CON(CH₃)₂ MW: 87.12 g/mol Boiling Pt: 166.1 Melting Pt: -20 Density: 0.937 g/mL Flash Pt: 70 °C (158 °F) |
MDL Number:
MFCD00008686 CAS Number: 127-19-5 EINECS: 204-826-4 UN: 2810 ADR: 6.1 III Merck Index: 13,03254 |
|
Specification Test Results
Assay | 99.5% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...