Silicic acid potassium salt, anhydrous
Supplier: BeanTown Chemical
224435-100G
224435-2KG
224435-500G
BT224435-100GEA
31.93
USD
BT224435-100G
BT224435-2KG
BT224435-500G
Silicic acid potassium salt, anhydrous
Silicic acid potassium salt
CAS: 1312-76-1; EC No: 215-199-1; MDL No: MFCD00145152
-48 Mesh Powder; Linear Formula: SiO2K2O (2.5:1 wt %); MW: 154.28
Density (g/mL): 2.4
- -48 Mesh Powder
Density:
2.4 g/mL |
MDL Number:
MFCD00145152 CAS Number: 1312-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...