(R)-(+)-N-Benzyl-α-methylbenzylamine ≥99%

Supplier: BeanTown Chemical

Synonyms: (+)-Benzylphenethylamine, (R)-N-Benzyl-1-phenylethanamine, (R)-(+)-N-Benzyl-1-phenylethylamine

224420-25G 224420-5G
BT224420-25GEA 124 USD
BT224420-25G BT224420-5G
(R)-(+)-N-Benzyl-α-methylbenzylamine ≥99%
(R)-(+)-N-Benzyl-α-methylbenzylamine

CAS: 38235-77-7; MDL No: MFCD00015010 Liquid; Molecular Formula: C15H17N ; MW: 211.30 Boiling Point: 171°/15 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.01; Refractive Index: 1.564; Optical Rotation: [α]20/D +38°, neat Air Sensitive


  • Optical rotation: [α]20/D +38°, neat
  • Refractive index: 1.564
  • Air Sensitive
  • Liquid

Formula: C₁₅H₁₇N
MW: 211.31 g/mol
Boiling Pt: 171 °C/15 mmHg
Density: 1.01 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00015010
CAS Number: 38235-77-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR